missing translation for 'onlineSavingsMsg'
Learn More

2-[(4-chlorophenyl)sulfonyl]ethanethioamide, 97%, Thermo Scientific™

Product Code. 10069481
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10069481 1 g Amber Glass Bottle 1g
10793663 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10069481 Supplier Thermo Scientific Maybridge Supplier No. SPB01724DA
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 59865-87-1
Molecular Formula C8H8ClNO2S2
Molecular Weight (g/mol) 249.73
MDL Number MFCD00052824
InChI Key OGQJCNYNVJXWBB-UHFFFAOYSA-N
Synonym 2-4-chlorophenyl sulfonyl ethanethioamide, 2-4-chlorobenzenesulfonyl ethanethioamide, maybridge1_008491, acmc-20ao70, 2-4-chlorophenylsulfonyl thioacetamide, 2-4-chlorophenyl sulfonylethanethioamide, 2-4-chlorophenyl sulfonyl thioacetamide, 4-chlorobenzene-1-sulfonyl ethanethioamide, 2-4-chloro-benzenesulfonyl-thioacetamide
PubChem CID 2743732
SMILES NC(=S)CS(=O)(=O)C1=CC=C(Cl)C=C1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula ClC6H4SO2CH2CSNH2
Quantity 1 g
IUPAC Name 2-(4-chlorophenyl)sulfonylethanethioamide
Formula Weight 249.74
Percent Purity 97%
Chemical Name or Material 2-[(4-Chlorophenyl)sulfonyl]ethanethioamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.