missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4-D-isopropyl ester 100 μg/mL in Cyclohexane, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | 2,4-D-iso-propyl ester |
| CAS | 94-11-1 |
| Analyte or Component Names | 2,4-D-iso-propyl ester |
| Shipping Condition | Room Temperature |
| Molecular Formula | C11 H12 Cl2 O3 |
| Concentration | 100 μg/mL |
| Certifications/Compliance | ISO/IEC 17025 |
| InChI Formula | InChI=1S/C11H12Cl2O3/c1-7(2)16-11(14)6-15-10-4-3-8(12)5-9(10)13/h3-5,7H,6H2,1-2H3 |
| Solution Type | Single Solution |
| SMILES | CC(C)OC(=O)COc1ccc(Cl)cc1Cl |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?