missing translation for 'onlineSavingsMsg'
Learn More

2,4-Di(chloromethyl)-1,3,5-trimethylbenzene, 97%, Thermo Scientific™

Product Code. 10283132
Change view
Click to view available options
Quantity:
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10283132 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10283132 Supplier Thermo Scientific Maybridge Supplier No. SB01371EA
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
This item is not returnable. View return policy

Specifications

CAS 1585-17-7
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C11H14Cl2
Linear Formula (CH3)3C6H(CH2Cl)2
MDL Number MFCD00013681
Quantity 10 g
Synonym 2,4-bis chloromethyl-1,3,5-trimethylbenzene, 2,4-bis chloromethyl mesitylene, benzene, 2,4-bis chloromethyl-1,3,5-trimethyl, 2,4-di chloromethyl-1,3,5-trimethylbenzene, 2,4-bis chloromethylmesitylene, acmc-209dhz, bis-chloromethyl mesitylene, bis-chloromethyl-mesitylene, jmndjdcloqrcjv-uhfffaoysa, 2,4-bis-chloromethyl mesitylene
InChI Key JMNDJDCLOQRCJV-UHFFFAOYSA-N
SMILES CC1=CC(C)=C(CCl)C(C)=C1CCl
IUPAC Name 2,4-bis(chloromethyl)-1,3,5-trimethylbenzene
Molecular Weight (g/mol) 217.13
PubChem CID 137095
Formula Weight 217.14
Percent Purity 97%
Chemical Name or Material 2,4-Di(chloromethyl)-1,3,5-trimethylbenzene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.