missing translation for 'onlineSavingsMsg'
Learn More

2,4-Di-tert-butylphenol, 97%

Product Code. p-3757464 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
2.5 kg
25 kg
Packaging:
Glass bottle
Metal Drum
Unit Size:
100g
2.5kg
25g
25kg
500g
5g
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10458510 5 g Glass bottle 5g
10732105 25 g Glass bottle 25g
10599043 100 g Glass bottle 100g
10553521 500 g Glass bottle 500g
10173590 2.5 kg Glass bottle 2.5kg
15254654 25 kg Metal Drum 25kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 10458510 Supplier Thermo Scientific Acros Supplier No. 160120050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 96-76-4 | C14H22O | 206.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96-76-4
MDL Number MFCD00008828
InChI Key ICKWICRCANNIBI-UHFFFAOYSA-N
Synonym 2,4-di-tert-butylphenol, 2,4-di-t-butylphenol, phenol, 2,4-bis 1,1-dimethylethyl, antioxidant no. 33, 1-hydroxy-2,4-di-tert-butylbenzene, phenol, 2,4-di-tert-butyl, 2,4-bis tert-butyl phenol, prodox 146, prodox 146a-85x, unii-fob94g6hzt
PubChem CID 7311
IUPAC Name 2,4-ditert-butylphenol
SMILES CC(C)(C)C1=CC(=C(C=C1)O)C(C)(C)C

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 54°C to 59°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C14H22O
Linear Formula [(CH3)3C]2C6H3OH
Quantity 5 g
Molecular Weight (g/mol) 206.329
Formula Weight 206.33
Percent Purity ≥96.0%
Physical Form Crystalline Solid
Chemical Name or Material 2, 4-Di-tert-butylphenol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.