missing translation for 'onlineSavingsMsg'
Learn More

2,4-Di-tert-butylphenol, TRC

Product Code. 17992135
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17992135 25 g 25g
17974455 100 g 100g
17991015 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17992135 Supplier Toronto Research Chemicals Supplier No. TRCD42947525G
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96-76-4
Molecular Formula C14 H22 O
Molecular Weight (g/mol) 206.32
Synonym Phenol, 2,4-di-tert-butyl- (8CI), 2,4-Bis(1,1-dimethylethyl)phenol, 2,4-Bis(tert-butyl)phenol, 2,4-Di-tert-butylhydroxybenzene, 2,4-Di-tert-butylphenol, Agidol 10, NSC 174502
IUPAC Name 2,4-ditert-butylphenol
SMILES CC(C)(C)c1ccc(O)c(c1)C(C)(C)C

Specifications

Quantity 25 g
InChI Formula InChI=1S/C14H22O/c1-13(2,3)10-7-8-12(15)11(9-10)14(4,5)6/h7-9,15H,1-6H3
Formula Weight 206.1671
Percent Purity >95%
Chemical Name or Material 2,4-Di-tert-butylphenol
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.