missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dibromo-1-fluorobenzene, tech. 90%, Thermo Scientific™

Product Code. 11440017
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11440017 25 g 25g
11430017 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11440017 Supplier Thermo Scientific Alfa Aesar Supplier No. B20955.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1435-53-6
Molecular Formula C6H3Br2F
Molecular Weight (g/mol) 253.90
MDL Number MFCD00000283
InChI Key MKHDDTWHDFVYDQ-UHFFFAOYSA-N
Synonym 2,4-dibromofluorobenzene, 1,3-dibromo-4-fluorobenzene, 1-fluoro-2,4-dibromobenzene, benzene, 2,4-dibromo-1-fluoro, pubchem3446, acmc-209cqz, 1,3-dibromo-5-fluoro, intermediates-zcf02097, ksc494g9j
PubChem CID 15034
IUPAC Name 2,4-dibromo-1-fluorobenzene
SMILES FC1=CC=C(Br)C=C1Br

Specifications

Density 2.047
Boiling Point 103°C (32 mmHg)
Flash Point 92°C (197°F)
Refractive Index 1.584
Quantity 25 g
Beilstein 1934822
Formula Weight 253.91
Percent Purity 90%
Grade Technical
Chemical Name or Material 2,4-Dibromo-1-fluorobenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.