missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dibromoanisole, 98%, Thermo Scientific™

Product Code. 10531652 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
50 g
Packaging:
Glass bottle
Unit Size:
50g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10531652 5 g Glass bottle 5g
10572251 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10531652 Supplier Thermo Scientific Acros Supplier No. 243230050
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 21702-84-1
Molecular Formula C7H6Br2O
Molecular Weight (g/mol) 265.92
MDL Number MFCD00000079
InChI Key XGXUGXPKRBQINS-UHFFFAOYSA-N
Synonym 2,4-dibromoanisole, benzene, 2,4-dibromo-1-methoxy, benzene,2,4-dibromo-1-methoxy, pubchem3847, acmc-209fnf, maybridge1_003667, chemwish ic06728, 2,4-dibromo-1-methoxy-benzene, xgxugxpkrbqins-uhfffaoysa, #
PubChem CID 27011
IUPAC Name 2,4-dibromo-1-methoxybenzene
SMILES COC1=C(C=C(C=C1)Br)Br

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 60.0°C to 64.0°C
Color Beige
Boiling Point 101.0°C to 106.0°C (1.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula Br2(C6H3)OCH3
Quantity 5 g
Formula Weight 265.92
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2, 4-Dibromoanisole, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.