missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dibromoanisole, 98+%, Thermo Scientific Chemicals

Product Code. 11413287
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11413287 25 g 25g
11423287 100 g 100g
11433287 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11413287 Supplier Thermo Scientific Alfa Aesar Supplier No. A16200.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21702-84-1
Molecular Formula C7H6Br2O
Molecular Weight (g/mol) 265.932
MDL Number MFCD00000079
InChI Key XGXUGXPKRBQINS-UHFFFAOYSA-N
PubChem CID 27011
IUPAC Name 2,4-dibromo-1-methoxybenzene
SMILES COC1=C(C=C(C=C1)Br)Br

Specifications

Melting Point 60°C to 64°C
Boiling Point 82°C to 88°C (0.3 mmHg)
Quantity 25 g
Beilstein 2045290
Synonym 2,4-dibromoanisole, benzene, 2,4-dibromo-1-methoxy, benzene,2,4-dibromo-1-methoxy, pubchem3847, acmc-209fnf, maybridge1_003667, chemwish ic06728, 2,4-dibromo-1-methoxy-benzene, xgxugxpkrbqins-uhfffaoysa, #
Formula Weight 265.93
Percent Purity ≥98%
Chemical Name or Material 2,4-Dibromoanisole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.