missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dibromophenol, 98%

Product Code. 11490997
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11490997

Brand: Thermo Scientific Alfa Aesar B23394.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 615-58-7 | C6H4Br2O | 251.905 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 615-58-7
Molecular Formula C6H4Br2O
Molecular Weight (g/mol) 251.905
MDL Number MFCD00002149
InChI Key FAXWFCTVSHEODL-UHFFFAOYSA-N
Synonym phenol, 2,4-dibromo, 2,4-dibromo-phenol, 2,4-dibromphenol, unii-ia75t5c9tg, 2,4-bis bromanyl phenol, ia75t5c9tg, phenol,4-dibromo, emery 9331, acmc-1bga9, 2,4-dibromophenol
PubChem CID 12005
ChEBI CHEBI:34238
IUPAC Name 2,4-dibromophenol
SMILES C1=CC(=C(C=C1Br)Br)O

Specifications

Melting Point 35°C to 38°C
Boiling Point 239°C
Flash Point >110°C (230°F)
Odor Phenol-like
Quantity 100 g
UN Number UN2811
Beilstein 1861291
Formula Weight 251.92
Percent Purity 98%
Chemical Name or Material 2,4-Dibromophenol
compliance-icons
Product Identifier
  • 2,4-Dibromophenol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H300-Fatal if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P330-Rinse mouth.
Supplemental information
  • MIXTURE LIST-Contains : 2,4-Dibromophenol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.