missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dibromophenol, 99%

Product Code. 10617561 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
5g
This item is not returnable. View return policy

Product Code. 10617561

Brand: Thermo Scientific Acros 406510050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 615-58-7 | C6H4Br2O | 251.91 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 615-58-7
Molecular Formula C6H4Br2O
Molecular Weight (g/mol) 251.91
MDL Number MFCD00002149
InChI Key FAXWFCTVSHEODL-UHFFFAOYSA-N
Synonym phenol, 2,4-dibromo, 2,4-dibromo-phenol, 2,4-dibromphenol, unii-ia75t5c9tg, 2,4-bis bromanyl phenol, ia75t5c9tg, phenol,4-dibromo, emery 9331, acmc-1bga9, 2,4-dibromophenol
PubChem CID 12005
ChEBI CHEBI:34238
IUPAC Name 2,4-dibromophenol
SMILES C1=CC(=C(C=C1Br)Br)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 36°C to 39°C
Color Beige or White to Yellow
Boiling Point 154°C (11.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula Br2C6H3OH
Quantity 5 g
Formula Weight 251.91
Percent Purity 99%
Physical Form Crystalline Mass, Chunks or Crystals and Powder
Chemical Name or Material 2, 4-Dibromophenol, 99%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.