missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichloro-1-iodobenzene, 97%, Thermo Scientific™

Product Code. 10767495
Change view
Click to view available options
Quantity:
50 g
100 g
Unit Size:
100g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10767495 50 g 50g
10566334 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10767495 Supplier Thermo Scientific Maybridge Supplier No. TL00185EE
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 29898-32-6
Molecular Formula C6H3Cl2I
Molecular Weight (g/mol) 272.894
InChI Key ZQKJCBDCOGLKCQ-UHFFFAOYSA-N
Synonym 1,3-dichloro-4-iodobenzene, 2,4-dichloroiodobenzene, benzene, 2,4-dichloro-1-iodo, 1-iodo-2,4-dichlorobenzene, pubchem3692, 2,4-dichlor-jodbenzol, acmc-209hbu, 1,3-dichloro4-iodobenzene, 1,3-dichloro 4-iodobenzene, 2,4-dichloro-1-iodo-benzene
PubChem CID 96864
IUPAC Name 2,4-dichloro-1-iodobenzene
SMILES C1=CC(=C(C=C1Cl)Cl)I

Specifications

CAS Min % 97.0
Density 2.0100g/mL
Assay Percent Range 97%
Quantity 50 g
Formula Weight 272.9
Percent Purity 97%
Chemical Name or Material 2, 4-Dichloro-1-iodobenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.