missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichloro-3-ethyl-6-nitrophenol, 98%, Thermo Scientific™

Product Code. 15214854 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
Product Code. Quantity Packaging unitSize
15214854 1 g Glass bottle 1g
15204854 5 g Glass bottle 5g
2 options
This item is not returnable. View return policy

Product Code. 15214854

Brand: Thermo Scientific Acros 450760010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Identificatori chimici

CAS 99817-36-4
Molecular Formula C8H6Cl2NO3
Molecular Weight (g/mol) 235.04
MDL Number MFCD00270764
InChI Key YTVCECQSAPGJBB-UHFFFAOYSA-M
Synonym 2,4-dichloro-3-ethyl-6-nitrophenol, 2,4-dichloro-3-ethyl-6-nitro-phenol, phenol, 2,4-dichloro-3-ethyl-6-nitro, 2-nitro-4,6-dichloro-5-ethylphenol, 2,4-dichloro-3-ethyl-6-nitrochloride, pubchem20442, acmc-209se5, ksc486m4b, 2,4-bis chloranyl-3-ethyl-6-nitro-phenol
PubChem CID 7020339
IUPAC Name 2,4-dichloro-3-ethyl-6-nitrobenzen-1-olate
SMILES CCC1=C(Cl)C=C(C([O-])=C1Cl)[N+]([O-])=O

Specifica

Melting Point 49.0°C to 51.0°C
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in methanol
Formula Weight 236.05
Percent Purity 98%
Chemical Name or Material 2, 4-Dichloro-3-ethyl-6-nitrophenol
compliance-icons compliance-icons compliance-icons
Identificativo prodotto
  • 2,4-Dichloro-3-ethyl-6-nitrophenol
Avvertenza
  • Danger
Categoria di pericolo
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
Indicazioni di pericolo
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H301-Toxic if swallowed.
Consigli di prudenza
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.