missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichloro-5-fluorobenzonitrile, 97%

Product Code. 11455810
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11455810 10 g 10g
11445810 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11455810 Supplier Thermo Scientific Alfa Aesar Supplier No. H26464.09
 Restricted Item
Estimated Shipment: 24-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 128593-93-1 | C7H2Cl2FN | 190.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 128593-93-1
Molecular Formula C7H2Cl2FN
Molecular Weight (g/mol) 190.00
MDL Number MFCD03840469
InChI Key XZBZWBDTFUFZSU-UHFFFAOYSA-N
PubChem CID 2782753
IUPAC Name 2,4-dichloro-5-fluorobenzonitrile
SMILES FC1=CC(C#N)=C(Cl)C=C1Cl

Specifications

Melting Point 44°C to 47°C
Quantity 10 g
UN Number UN3439
Synonym 2,4-dichloro-5-fluoro-benzonitrile, pubchem1566, acmc-1bxxv, 2,4-dichloro-5-fluoro benzonitrile, benzonitrile,2,4-dichloro-5-fluoro, benzonitrile, 2,4-dichloro-5-fluoro, 2,4-dichloro-5-fluorobenzonitrile, 2,4-dichloro-5-fluorobenzenecarbonitrile, 2,4-bis chloranyl-5-fluoranyl-benzenecarbonitrile
Formula Weight 190
Percent Purity 97%
Chemical Name or Material 2,4-Dichloro-5-fluorobenzonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.