missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichloro-5-sulfamoylbenzoic acid, 98%, Thermo Scientific Chemicals

Product Code. 11430438
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11430438 50 g 50g
11440438 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11430438 Supplier Thermo Scientific Alfa Aesar Supplier No. B24589.18
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2736-23-4 | Molecular Formula: C7H5Cl2NO4S | Molecular Weight: 270.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2736-23-4
Molecular Formula C7H5Cl2NO4S
Molecular Weight (g/mol) 270.08
MDL Number MFCD00007931
InChI Key ZSHHRBYVHTVRFK-UHFFFAOYSA-N
Synonym lasamide, 5-aminosulfonyl-2,4-dichlorobenzoic acid, 2,4-dichloro-5-sulphamoylbenzoic acid, 2,4-dichloro-5-sulfamoyl-benzoic acid, unii-leg53tf0sn, 2,4-dichloro-5-sulfamylbenzoic acid, benzoic acid, 5-aminosulfonyl-2,4-dichloro, leg53tf0sn, 3-sulfamoyl-4,6-dichlorobenzoic acid, benzoic acid, 2,4-dichloro-5-sulfamoyl
PubChem CID 17655
IUPAC Name 2,4-dichloro-5-sulfamoylbenzoic acid
SMILES C1=C(C(=CC(=C1S(=O)(=O)N)Cl)Cl)C(=O)O

Specifications

Melting Point 230°C to 232°C (decomposition)
Quantity 50 g
Formula Weight 270.09
Percent Purity 98%
Chemical Name or Material 2,4-Dichloro-5-sulfamoylbenzoic acid

RUO – Research Use Only

Product Title
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.