missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichloro-6-methylaniline, 97%

Product Code. 11414314
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11414314 5 g 5g
11424314 25 g 25g
11434314 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11414314 Supplier Thermo Scientific Alfa Aesar Supplier No. A12591.06
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 30273-00-8 | C7H7Cl2N | 176.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 30273-00-8
Molecular Formula C7H7Cl2N
Molecular Weight (g/mol) 176.04
MDL Number MFCD00044074
InChI Key UAISVUGQLKXPFF-UHFFFAOYSA-N
Synonym 4,6-dichloro-2-methylaniline, benzenamine, 2,4-dichloro-6-methyl, acmc-20aodo, 4,6-dichloro-o-toluidine, 2-amino-3,5-dichlorotoluene, 2,4-dichloro-6-methylphenylamine, 4,6-dichloro-2-methylphenylamine, 2,4-bis chloranyl-6-methyl-aniline, 2,4-dichloro-6-methylaniline
PubChem CID 1268287
IUPAC Name 2,4-dichloro-6-methylaniline
SMILES CC1=CC(=CC(=C1N)Cl)Cl

Specifications

Melting Point 43°C to 45°C
Boiling Point 82°C to 84°C (0.1 mmHg)
Quantity 5 g
Beilstein 2208221
Formula Weight 176.05
Percent Purity 97%
Chemical Name or Material 2,4-Dichloro-6-methylaniline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.