missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichloro-6-methylbenzylamine, 98+%, Thermo Scientific™

Product Code. 11332897
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11332897 5 g 5g
11342897 250 mg 250mg
11322897 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11332897 Supplier Thermo Scientific Alfa Aesar Supplier No. L16108.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 150517-76-3
Molecular Formula C8H9Cl2N
Molecular Weight (g/mol) 190.07
MDL Number MFCD00052913
InChI Key QISTWSMZJRTDCS-UHFFFAOYSA-N
Synonym 2,4-dichloro-6-methylbenzylamine, 2,4-dichloro-6-methylphenyl methanamine, 2,4-dichloro-6-methylphenyl methylamine, benzenemethanamine,2,4-dichloro-6-methyl, benzenemethanamine, 2,4-dichloro-6-methyl, 2,4-dichloro-6-methylbenzyl amine hydrochloride, 4,6-dichloro-2-methylbenzylamine, acmc-1c3je, 2-aminomethyl-3,5-dichlorotoluene, 1-2,4-dichloro-6-methylphenyl methanamine
PubChem CID 2800971
SMILES CC1=CC(Cl)=CC(Cl)=C1CN

Specifications

Melting Point 52°C to 56°C
Quantity 5 g
UN Number UN3259
Beilstein 8615227
Sensitivity Air Sensitive
IUPAC Name 1-(2,4-dichloro-6-methylphenyl)methanamine
Formula Weight 190.07
Percent Purity ≥98%
Chemical Name or Material 2,4-Dichloro-6-methylbenzylamine
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.