missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichloro-6-nitrophenol, 98+%

Product Code. 15431779
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15431779 100 g 100g
15421779 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15431779 Supplier Thermo Scientific Alfa Aesar Supplier No. H64524.22
 Restricted Item
Estimated Shipment: 21-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 609-89-2 | C6H3Cl2NO3 | 207.994 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 609-89-2
Molecular Formula C6H3Cl2NO3
Molecular Weight (g/mol) 207.994
MDL Number MFCD00007101
InChI Key LYPMXMBQPXPNIQ-UHFFFAOYSA-N
Synonym 2,4-dichloro-6-nitrophenol, phenol, 2,4-dichloro-6-nitro, 2,4-dichlor-6-nitrofenol, 4,6-dichloro-2-nitrophenol, 2,4-dichlor-6-nitrofenol czech, pubchem22918, acmc-1aunw, wln: wnr bq cg eg, 2-nitro-4,6-dichlorophenol
PubChem CID 11871
SMILES C1=C(C=C(C(=C1Cl)O)[N+](=O)[O-])Cl

Specifications

Melting Point 118°C to 120°C
Color Yellow
Quantity 100 g
UN Number UN2811
Beilstein 2050081
Formula Weight 207.99
Percent Purity ≥98%
Physical Form Powder
Chemical Name or Material 2,4-Dichloro-6-nitrophenol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.