missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichloroanisole, 99%

Product Code. 11450511
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11450511 5 g 5g
11460511 25 g 25g
11470511 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11450511 Supplier Thermo Scientific Alfa Aesar Supplier No. B20042.06
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 553-82-2 | C7H6Cl2O | 177.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 553-82-2
Molecular Formula C7H6Cl2O
Molecular Weight (g/mol) 177.02
MDL Number MFCD00044772
InChI Key CICQUFBZCADHHX-UHFFFAOYSA-N
Synonym 2,4-dichloroanisole, benzene, 2,4-dichloro-1-methoxy, 1,5-dichloro-2-methoxybenzene, 2,4-dichloro-1-methoxy-benzene, anisole, 2,4-dichloro, 2,4-dichloromethoxybenzene, anisole,4-dichloro, acmc-209lmo, ksc273i7j
PubChem CID 11119
IUPAC Name 2,4-dichloro-1-methoxybenzene
SMILES COC1=CC=C(Cl)C=C1Cl

Specifications

Melting Point 24°C to 27°C
Density 1.288
Boiling Point 125°C (10 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.5615
Quantity 5 g
Beilstein 1940358
Formula Weight 177.03
Percent Purity 99%
Chemical Name or Material 2,4-Dichloroanisole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.