missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichlorobenzoyl chloride, 98%

Product Code. 10756201 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
100 mL
500 mL
Packaging:
Glass bottle
Unit Size:
100mL
500mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10756201 500 mL Glass bottle 500mL
10275100 5 mL Glass bottle 5mL
10103680 100 mL Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10756201 Supplier Thermo Scientific Acros Supplier No. 150225000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 89-75-8 | C7H3Cl3O | 209.45 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89-75-8
Molecular Formula C7H3Cl3O
Molecular Weight (g/mol) 209.45
MDL Number MFCD00000661
InChI Key CEOCVKWBUWKBKA-UHFFFAOYSA-N
Synonym benzoyl chloride, 2,4-dichloro, 2,4-dichlorobenzoylchloride, benzoylchloride, 2,4-dichloro, ccris 8616, acmc-1cuf7, dsstox_cid_31176, dsstox_gsid_52603, 2,4-dichloro benzoylchloride, 2.4-dichlorobenzoyl chloride, ksc450c2p
PubChem CID 66645
IUPAC Name 2,4-dichlorobenzoyl chloride
SMILES ClC(=O)C1=CC=C(Cl)C=C1Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 21°C
Color Colorless
Density 1.4900g/mL
Boiling Point 150°C (34.0 mmHg)
Flash Point 137°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula Cl2C6H3COCl
Refractive Index 1.5915 to 1.5937
Quantity 500 mL
Beilstein 09, 342
Specific Gravity 1.49
Solubility Information Solubility in water: may decompose. Other solubilities: soluble in alcohol,ether and acetone,slightly soluble in heptane
Formula Weight 209.46
Percent Purity 98%
Physical Form Liquid After Melting
Chemical Name or Material 2, 4-Dichlorobenzoyl chloride, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 2,4-Dichlorobenzoyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • EUH014-Reacts violently with water.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.