missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichlorobenzyl alcohol, 99%

Product Code. 11450963
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11450963

Brand: Thermo Scientific Alfa Aesar A13338.36

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1777-82-8 | C7H6Cl2O | 177.024 g/mol

2,4-Dichlorobenzyl alcohol is an antiseptic found in commercially available throat lozenges. Also used deactivate Respiratory syncytial virus and SARS-Cov.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4-Dichlorobenzyl alcohol is an antiseptic found in commercially available throat lozenges. Also used deactivate Respiratory syncytial virus and SARS-Cov.

Solubility
Soluble in water, methanol and chloroform.

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1777-82-8
Molecular Formula C7H6Cl2O
Molecular Weight (g/mol) 177.024
MDL Number MFCD00004606
InChI Key DBHODFSFBXJZNY-UHFFFAOYSA-N
Synonym 2,4-dichlorobenzyl alcohol, 2,4-dichlorophenyl methanol, dybenal, 2,4-dichlorobenzenemethanol, rapidosept, myacide sp, benzenemethanol, 2,4-dichloro, benzyl alcohol, 2,4-dichloro, unii-1nkx3648j9, 2,4-dichlorobenzylalcohol
PubChem CID 15684
ChEBI CHEBI:48220
IUPAC Name (2,4-dichlorophenyl)methanol
SMILES C1=CC(=C(C=C1Cl)Cl)CO

Specifications

Melting Point 57°C to 61°C
Flash Point >110°C (230°F)
Quantity 500 g
Beilstein 1448652
Merck Index 14,3060
Solubility Information Soluble in water,methanol and chloroform.
Formula Weight 177.03
Percent Purity 99%
Chemical Name or Material 2,4-Dichlorobenzyl alcohol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.