missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichlorobenzyl chloride, 98%

Product Code. 11444090
Click to view available options
Quantity:
50 g
500 g
1000 g
Unit Size:
1000g
500g
50g
This item is not returnable. View return policy

Product Code. 11444090

Brand: Thermo Scientific Alfa Aesar A11435.36

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 94-99-5 | C7H5Cl3 | 195.47 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 94-99-5
Molecular Formula C7H5Cl3
Molecular Weight (g/mol) 195.47
MDL Number MFCD00000895
InChI Key IRSVDHPYXFLLDS-UHFFFAOYSA-N
Synonym 2,4-dichlorobenzyl chloride, 2,4-dichloro-1-chloromethyl benzene, 2,4-dichlorobenzylchloride, alpha,2,4-trichlorotoluene, benzene, 2,4-dichloro-1-chloromethyl, 2,4-dichloro-1-chloromethyl-benzene, alpha2,4-trichlorotoluene, unii-85e96p2s3s, 2,4-dcbc, toluene, .alpha.2,4-trichloro
PubChem CID 7212
IUPAC Name 2,4-dichloro-1-(chloromethyl)benzene
SMILES ClCC1=CC=C(Cl)C=C1Cl

Specifications

Melting Point -3°C
Density 1.408
Boiling Point 248°C
Flash Point 190°C (374°F)
Odor Pungent
Refractive Index 1.576
Quantity 500 g
UN Number UN3265
Beilstein 387220
Formula Weight 195.48
Percent Purity 98%
Chemical Name or Material 2,4-Dichlorobenzyl chloride
Show More Show Less
compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.