missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichlorobenzyl chloride, 99%

Product Code. 10192990 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 mL
100 mL
500 mL
Packaging:
Glass bottle
Unit Size:
100mL
500mL
5mL
This item is not returnable. View return policy

Product Code. 10192990

Brand: Thermo Scientific Acros 134390050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 94-99-5 | C7H5Cl3 | 195.47 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 94-99-5
Molecular Formula C7H5Cl3
Molecular Weight (g/mol) 195.47
MDL Number MFCD00000895
InChI Key IRSVDHPYXFLLDS-UHFFFAOYSA-N
Synonym 2,4-dichlorobenzyl chloride, 2,4-dichloro-1-chloromethyl benzene, 2,4-dichlorobenzylchloride, alpha,2,4-trichlorotoluene, benzene, 2,4-dichloro-1-chloromethyl, 2,4-dichloro-1-chloromethyl-benzene, alpha2,4-trichlorotoluene, unii-85e96p2s3s, 2,4-dcbc, toluene, .alpha.2,4-trichloro
PubChem CID 7212
IUPAC Name 2,4-dichloro-1-(chloromethyl)benzene
SMILES ClCC1=CC=C(Cl)C=C1Cl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -2.7°C
Color Colorless to Yellow
Density 1.4000g/mL
Boiling Point 248.0°C
Flash Point 190°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula Cl2C6H3CH2Cl
Refractive Index 1.5750 to 1.5770
Quantity 5 mL
Specific Gravity 1.4
Solubility Information Solubility in water: hydrolysis by water. Other solubilities: soluble in alcohol,ether and acetone
Viscosity 2.6 mm2/s (20°C)
Formula Weight 195.48
Percent Purity 99%
Physical Form Oily Liquid
Chemical Name or Material 2, 4-Dichlorobenzyl chloride, 99%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2,4-Dichlorobenzyl chloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.