missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichlorobenzylamine, 98%

Product Code. 10132760 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 mL
50 mL
Packaging:
Glass bottle
Unit Size:
10mL
50mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10132760 10 mL Glass bottle 10mL
10294420 50 mL Glass bottle 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10132760 Supplier Thermo Scientific Acros Supplier No. 113230100
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 95-00-1 | C7H7Cl2N | 176.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-00-1
Molecular Formula C7H7Cl2N
Molecular Weight (g/mol) 176.04
MDL Number MFCD00008109
InChI Key SJUKJZSTBBSGHF-UHFFFAOYSA-N
Synonym 2,4-dichlorobenzylamine, 2,4-dichlorophenyl methanamine, benzenemethanamine, 2,4-dichloro, benzylamine, 2,4-dichloro, 2,4-dichlorobenzyl amine, 1-2,4-dichlorophenyl methanamine, 2,4-dichloro-benzylamine, 2,4-dichlorophenyl methylamine, o,p-dichlorobenzylamine, acmc-209rur
PubChem CID 1485
IUPAC Name (2,4-dichlorophenyl)methanamine
SMILES NCC1=CC=C(Cl)C=C1Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless
Density 1.3200g/mL
Boiling Point 258.0°C to 260.0°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula Cl2C6H3CH2NH2
Refractive Index 1.5765 to 1.5785
Quantity 10 mL
Specific Gravity 1.32
Formula Weight 176.04
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2, 4-Dichlorobenzylamine, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 2,4-Dichlorobenzylamine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.