missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichlorophenol 100 μg/mL in Methanol, Dr. Ehrenstorfer

Product Code. 30312075
Change view
Click to view available options
Quantity:
5 x 1 mL
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30312075 5 x 1 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30312075 Supplier Dr. Ehrenstorfer Supplier No. DREGS09010065ME

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,4-Dichlorophenol
CAS 120-83-2
Analyte or Component Names 2,4-Dichlorophenol
Shipping Condition +20°C
Molecular Formula C6 H4 Cl2 O
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
Synonym 2,4-Dichlorophenol, 2,4-Dichlorphenol, Phenol, 2,4-dichloro-, 1,3-Dichloro-4-hydroxybenzene, 2,4-DCP, 2,4-Dichlorophenic acid, 4,6-Dichlorophenol, DCP, NSC 2879
InChI Formula InChI=1S/C6H4Cl2O/c7-4-1-2-6(9)5(8)3-4/h1-3,9H
SMILES Oc1ccc(Cl)cc1Cl
IUPAC Name 2,4-dichlorophenol
Solvent or Matrix Methanol
Molecular Weight (g/mol) 163
Quantity 5 x 1 mL
Formula Weight 161.9639 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.