missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichlorophenoxyacetic acid, 98.0%, MP Biomedicals™

Product Code. 11447900 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11447900 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11447900 Supplier MP Biomedicals Supplier No. 0210155380
 Restricted Item
Estimated Shipment: 23-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 94-75-7
Color Orange-Tan to White
Density 0.66g/cm3 (bulk density)
Assay Percent Range 98.0%
Molecular Formula C8H6Cl2O3
Quantity 100 g
Synonym 2,4-dichlorophenoxyacetic acid, 2-2,4-dichlorophenoxy acetic acid, 2,4-dichlorophenoxy acetic acid, hedonal, rhodia, agrotect, fernesta, fernimine, netagrone, tributon
InChI Key OVSKIKFHRZPJSS-UHFFFAOYSA-N
SMILES C1=CC(=C(C=C1Cl)Cl)OCC(=O)O
IUPAC Name 2-(2,4-dichlorophenoxy)acetic acid
Molecular Weight (g/mol) 221.033
PubChem CID 1486
ChEBI CHEBI:28854
Formula Weight 221.04
Physical Form Powder
Chemical Name or Material 2, 4-Dichlorophenoxyacetic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.