missing translation for 'onlineSavingsMsg'
Learn More

2,4-Difluoro-1-iodobenzene, 99%

Product Code. 11499712
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11499712 10 g 10g
11409722 50 g 50g
11419722 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11499712 Supplier Thermo Scientific Alfa Aesar Supplier No. A10798.09
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 2265-93-2 | C6H3F2I | 239.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2265-93-2
Molecular Formula C6H3F2I
Molecular Weight (g/mol) 239.99
MDL Number MFCD00040151
InChI Key YKLDMAPEGQYZRT-UHFFFAOYSA-N
Synonym 2,4-difluoroiodobenzene, 1,3-difluoro-4-iodobenzene, 2,4-difluoro-1-iodobenze, benzene, 2,4-difluoro-1-iodo, 2,4-difluoro iodobenzene, 2,4-difluoro-1-iodo-benzene, pubchem3463, 2,4-difluoro-iodobenzene, acmc-1cq0v, intermediates-zcf02607
PubChem CID 137519
IUPAC Name 2,4-difluoro-1-iodobenzene
SMILES FC1=CC=C(I)C(F)=C1

Specifications

Density 2.006
Boiling Point 175°C to 176°C
Flash Point 67°C (152°F)
Refractive Index 1.558
Quantity 10 g
Beilstein 2081078
Sensitivity Light sensitive
Formula Weight 239.99
Percent Purity 99%
Chemical Name or Material 2,4-Difluoro-1-iodobenzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.