missing translation for 'onlineSavingsMsg'
Learn More

2,4-Difluoro-3-methoxybenzoic acid, 98+%

Product Code. p-7028337
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11401090

Brand: Thermo Scientific Alfa Aesar B25597.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 178974-97-5 | C8H6F2O3 | 188.13 g/mol

2,4-Difluoro-3-methoxybenzoic acid is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4-Difluoro-3-methoxybenzoic acid is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 178974-97-5
Molecular Formula C8H6F2O3
Molecular Weight (g/mol) 188.13
MDL Number MFCD02258905
InChI Key KWLIGHXPUYUTBH-UHFFFAOYSA-N
Synonym 2,4-difluoro-3-methoxy-benzoic acid, 2,4-difluoro-3-methoxylbenzoic acid, 2,4-difluoro-3-methoxy benzoic acid, pubchem4941, acmc-1cabj, 3-methoxy-2,4-difluorobenzoic acid, benzoic acid,2,4-difluoro-3-methoxy
PubChem CID 3780555
IUPAC Name 2,4-difluoro-3-methoxybenzoic acid
SMILES COC1=C(F)C=CC(C(O)=O)=C1F

Specifications

Melting Point 193°C to 195°C
Quantity 1 g
Solubility Information Insoluble in water.
Formula Weight 188.13
Percent Purity ≥98%
Chemical Name or Material 2,4-Difluoro-3-methoxybenzoic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.