missing translation for 'onlineSavingsMsg'
Learn More

2,4-Difluoro-3-methylbenzylamine, 98%, Thermo Scientific™

Product Code. p-7031104
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15454737 250 mg 250mg
15434737 1 g 1g
15444737 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15454737 Supplier Thermo Scientific Alfa Aesar Supplier No. H32060.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 847502-90-3
Molecular Formula C8H9F2N
Molecular Weight (g/mol) 157.164
MDL Number MFCD08275293
InChI Key LPMVSOQQHWTDRH-UHFFFAOYSA-N
Synonym 2,4-difluoro-3-methylbenzylamine, 1-2,4-difluoro-3-methylphenyl methanamine, 2,4-difluoro-3-methylphenyl methanamine, benzenemethanamine,2,4-difluoro-3-methyl
PubChem CID 11469203
IUPAC Name (2,4-difluoro-3-methylphenyl)methanamine
SMILES CC1=C(C=CC(=C1F)CN)F

Specifications

Boiling Point 42°C (1 mmHg)
Refractive Index 1.495
Quantity 250 mg
UN Number UN2735
Sensitivity Air Sensitive
Formula Weight 157.16
Percent Purity 98%
Chemical Name or Material 2,4-Difluoro-3-methylbenzylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.