missing translation for 'onlineSavingsMsg'
Learn More

2,4-Difluorobenzene-1-carbothioamide, ≥97%, Maybridge

Product Code. 10057392
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 10057392

Brand: Thermo Scientific Maybridge SPB02253DA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 175276-92-3
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C7H5F2NS
MDL Number MFCD00085011
Quantity 1 g
Synonym 2,4-difluorobenzene-1-carbothioamide, 2,4-difluorothiobenzamide, 2,4-difluorobenzothioamide, acmc-1c1ty, 2,4-difluoro-thiobenzamide, 2,4-difluorobenzene carbothioamide, 2,4-bis fluoranyl benzenecarbothioamide, amino 2,4-difluorophenyl methane-1-thione, benzenecarbothioamide, 2,4-difluoro-9ci
InChI Key MOHAZBCWHUCIEX-UHFFFAOYSA-N
SMILES C1=CC(=C(C=C1F)F)C(=S)N
IUPAC Name 2,4-difluorobenzenecarbothioamide
Molecular Weight (g/mol) 173.181
PubChem CID 2781868
Formula Weight 173.19
Percent Purity 97%
Chemical Name or Material 2, 4-Difluorobenzene-1-carbothioamide
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.