missing translation for 'onlineSavingsMsg'
Learn More

2,4-Difluorobenzoic acid, 98%

Product Code. 11425423
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11425423

Brand: Thermo Scientific Alfa Aesar A13403.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1583-58-0 | C7H4F2O2 | 158.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1583-58-0
Molecular Formula C7H4F2O2
Molecular Weight (g/mol) 158.10
MDL Number MFCD00011670
InChI Key NJYBIFYEWYWYAN-UHFFFAOYSA-N
Synonym 2,4-dfba, benzoic acid, 2,4-difluoro, 2,4-difluorobenzoicacid, 2,4-difluoro-benzoic acid, zlchem 472, pubchem1336, acmc-1brgf, 2,4-difluorbenzoic acid, 2,4-difluorobezoic acid, 2 4-difluorobenzoic acid
PubChem CID 74102
IUPAC Name 2,4-difluorobenzoic acid
SMILES OC(=O)C1=CC=C(F)C=C1F

Specifications

Melting Point 186°C to 190°C
Odor Odorless
Quantity 25 g
Beilstein 973355
Formula Weight 158.11
Percent Purity 98%
Chemical Name or Material 2,4-Difluorobenzoic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.