missing translation for 'onlineSavingsMsg'
Learn More

2,4'-Difluorobenzophenone, 97%, Thermo Scientific™

Product Code. 15488919
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15488919 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15488919 Supplier Thermo Scientific Alfa Aesar Supplier No. L10306.06
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 342-25-6
Melting Point 22°C to 24°C
Density 1.244
Boiling Point 176°C to 178°C (16 mmHg)
Flash Point >110°C (230°F)
Molecular Formula C13H8F2O
Refractive Index 1.5689
MDL Number MFCD00000319
Quantity 5 g
Beilstein 3268520
Synonym 2,4'-difluorobenzophenone, 2-fluorophenyl 4-fluorophenyl methanone, 2',4-difluorobenzophenone, 2-fluorophenyl-4-fluorophenyl methanone, di2-fluorophenyl ketone, fr bvr df, pubchem7479, acmc-1clee, 2,4;-difluorobenzophenone, 2,4/'-difluorobenzophenone
InChI Key LKFIWRPOVFNPKR-UHFFFAOYSA-N
SMILES C1=CC=C(C(=C1)C(=O)C2=CC=C(C=C2)F)F
IUPAC Name (2-fluorophenyl)-(4-fluorophenyl)methanone
Molecular Weight (g/mol) 218.203
PubChem CID 67651
Formula Weight 218.2
Percent Purity 97%
Chemical Name or Material 2,4'-Difluorobenzophenone
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.