Product Code. 10044772
Shop All Thermo Scientific Chemicals Products
Click to view available options
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
Quantity
Packaging
Unit Size
10044772
5 g
Glass bottle
5g
10359500
1 g
Glass bottle
1g
10177993
25 g
Glass bottle
25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code.
10044772
Supplier
Thermo Scientific Acros
Supplier No.
305660050
Estimated Shipment: 27-11-2026Log in to see stock.
missing translation for 'validMainframeMsg'
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
CAS: 72235-52-0 | C7 H7 F2 N | 143.14 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
CAS
72235-52-0
Molecular Formula
C7 H7 F2 N
Molecular Weight (g/mol)
143.14
MDL Number
MFCD00010142
InChI Key
QDZZDVQGBKTLHV-UHFFFAOYSA-N
Synonym
2,4-difluorobenzylamine, 2,4-difluorophenyl methanamine, 2,4-difluorobenzyl amine, benzenemethanamine, 2,4-difluoro, 1-2,4-difluorophenyl methanamine, 2,4-difluorophenyl methylamine, n-difluorobenzylamine, pubchem4399, 2,4-difluorbenzylamine, acmc-209oma
PubChem CID
2733244
IUPAC Name
(2,4-difluorophenyl)methanamine
SMILES
C1=CC(=C(C=C1F)F)CN
Specifications
CAS Min %
97.5
CAS Max %
100.0
Color
Colorless to Yellow
Density
1.2000g/mL
Boiling Point
82°C to 84°C (15.0 mmHg)
Flash Point
68°C
Infrared Spectrum
Authentic
Assay Percent Range
97.5% min. (GC)
Packaging
Glass bottle
Linear Formula
F2 C6 H3 CH2 NH2
Refractive Index
1.4900 to 1.493
Quantity
5 g
Specific Gravity
1.2
Formula Weight
143.14
Percent Purity
98%
Physical Form
Liquid
Chemical Name or Material
2, 4-Difluorobenzylamine
Show More
Show Less
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our
Privacy Policy .
Spot an opportunity for improvement?