missing translation for 'onlineSavingsMsg'
Learn More

2,4-Difluorobenzylamine, 98%

Product Code. 10359500 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10359500 1 g Glass bottle 1g
10044772 5 g Glass bottle 5g
10177993 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 10359500 Supplier Thermo Scientific Acros Supplier No. 305660010
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 72235-52-0 | C7H7F2N | 143.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72235-52-0
Molecular Formula C7H7F2N
Molecular Weight (g/mol) 143.14
MDL Number MFCD00010142
InChI Key QDZZDVQGBKTLHV-UHFFFAOYSA-N
Synonym 2,4-difluorobenzylamine, 2,4-difluorophenyl methanamine, 2,4-difluorobenzyl amine, benzenemethanamine, 2,4-difluoro, 1-2,4-difluorophenyl methanamine, 2,4-difluorophenyl methylamine, n-difluorobenzylamine, pubchem4399, 2,4-difluorbenzylamine, acmc-209oma
PubChem CID 2733244
IUPAC Name (2,4-difluorophenyl)methanamine
SMILES C1=CC(=C(C=C1F)F)CN

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.2000g/mL
Boiling Point 82°C to 84°C (15.0 mmHg)
Flash Point 68°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula F2C6H3CH2NH2
Refractive Index 1.4900 to 1.493
Quantity 1 g
Specific Gravity 1.2
Formula Weight 143.14
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2, 4-Difluorobenzylamine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.