missing translation for 'onlineSavingsMsg'
Learn More

2,4-Difluorobenzylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific™

Product Code. 15495305
Change view
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15495305 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15495305 Supplier Thermo Scientific Alfa Aesar Supplier No. H58036.AD
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2,4-Difluorobenzylzinc bromide is used as pharmaceutical intermediates.

Solubility
Reacts with water.

Notes
Store at 4°C. Protect from heat. Store away from air. Air Sensitive. Do not store toge
TRUSTED_SUSTAINABILITY

Specifications

CAS 307496-26-0
Density 0.995
Molecular Formula C7H5BrF2Zn
MDL Number MFCD01311423
Quantity 50 mL
UN Number UN3399
Sensitivity Air sensitive; May form a precipitate
Synonym 2,4-difluorobenzylzinc bromide, bromo 2,4-difluorophenyl methyl zinc, 2,4-difluorobenzylzinc bromide 0.5m solution in thf, 2,4-difluorobenzylzinc bromide 0.5 m in tetrahydrofuran, 2,4-difluorobenzylzinc bromide solution, 0.5 m in thf, 2,4-difluorobenzylzinc bromide, 0.5m in thf, packaged under argon in resealable chemseal bottles
Solubility Information Reacts with water.
InChI Key WPJAHFYOACWPNA-UHFFFAOYSA-M
SMILES [CH2-]C1=C(C=C(C=C1)F)F.[Zn+]Br
IUPAC Name bromozinc(1+);2,4-difluoro-1-methanidylbenzene
Molecular Weight (g/mol) 272.398
PubChem CID 2778322
Formula Weight 272.4
Chemical Name or Material 2,4-Difluorobenzylzinc bromide, Packaged under Argon in resealable ChemSeal™ bottles
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.