missing translation for 'onlineSavingsMsg'
Learn More

2,4-Difluorophenol, 98+%

Product Code. 11497936
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 11497936

Brand: Thermo Scientific Alfa Aesar A14044.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 367-27-1 | C6H4F2O | 130.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 367-27-1
Molecular Formula C6H4F2O
Molecular Weight (g/mol) 130.09
MDL Number MFCD00009715
InChI Key NVWVWEWVLBKPSM-UHFFFAOYSA-N
PubChem CID 123051
IUPAC Name 2,4-difluorophenol
SMILES OC1=CC=C(F)C=C1F

Specifications

Melting Point 21°C to 24°C
Boiling Point 52°C to 54°C (14 mmHg)
Flash Point 56°C (132°F)
Odor Phenol-like
Quantity 10 g
UN Number UN2921
Beilstein 1934803
Synonym phenol, 2,4-difluoro, 2.4-difluorophenol, 2,4difluorophenol, 2,4-diflurophenol, 2 4-difluorophenol, 2,4 difluorophenol, 2, 4 difluorophenol, 2,4 difluoro phenol, 2,4-di-fluorophenol, 2,4-difluoro phenol
Formula Weight 130.09
Percent Purity ≥98%
Chemical Name or Material 2,4-Difluorophenol
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.