missing translation for 'onlineSavingsMsg'
Learn More

2,4-Difluorophenyl isocyanate, 98+%

Product Code. 11499593
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11499593 5 g 5g
11409603 25 g 25g
11419603 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11499593 Supplier Thermo Scientific Alfa Aesar Supplier No. A12415.06
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 59025-55-7 | C7H3F2NO | 155.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59025-55-7
Molecular Formula C7H3F2NO
Molecular Weight (g/mol) 155.10
MDL Number MFCD00001997
InChI Key HNENEALJPWJWJY-UHFFFAOYSA-N
Synonym 2,4-difluorophenyl isocyanate, 2,4-difluorophenylisocyanate, benzene, 2,4-difluoro-1-isocyanato, isocyanic acid 2,4-difluorophenyl ester, 2,4-difluoro-1-isocyanato-benzene, 2,4-difluoroisocyanatobenzene, timtec-bb sbb006537, pubchem4453, 2,4-diflurophenylisocyanate, acmc-1b06k
PubChem CID 593258
IUPAC Name 2,4-difluoro-1-isocyanatobenzene
SMILES FC1=CC=C(N=C=O)C(F)=C1

Specifications

Density 1.31
Boiling Point 64°C to 66°C (10 mmHg)
Flash Point 50°C (122°F)
Refractive Index 1.492
Quantity 5 g
UN Number UN3080
Beilstein 2831577
Sensitivity Moisture sensitive
Formula Weight 155.11
Percent Purity ≥98%
Chemical Name or Material 2,4-Difluorophenyl isocyanate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.