missing translation for 'onlineSavingsMsg'
Learn More

2,4-Difluorophenyl isothiocyanate, 96%

Product Code. 11479646
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11479646

Brand: Thermo Scientific Alfa Aesar B20204.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 141106-52-7 | C7H3F2NS | 171.165 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141106-52-7
Molecular Formula C7H3F2NS
Molecular Weight (g/mol) 171.165
MDL Number MFCD00041044
InChI Key ABGGPKIFVAIRGU-UHFFFAOYSA-N
Synonym 2,4-difluorophenyl isothiocyanate, 2,4-difluoro isothiocyanato benzene, 2,4-difluoro-1-isothiocyanato-benzene, 2,4-difluorophenylisothiocyanate, isothiocyanic acid 2,4-difluorophenyl ester, benzene, 2,4-difluoro-1-isothiocyanato, benzene, 2,4-difluoro-1-isothiocyanato-9ci, 2,4-difluorobenzenisothiocyanate, pubchem4451, acmc-209clq
PubChem CID 518879
IUPAC Name 2,4-difluoro-1-isothiocyanatobenzene
SMILES C1=CC(=C(C=C1F)F)N=C=S

Specifications

Density 1.349
Boiling Point 98°C to 99°C (30 mmHg)
Flash Point 92°C (197°F)
Refractive Index 1.597
Quantity 1 g
UN Number UN2922
Beilstein 6919744
Sensitivity Moisture sensitive
Formula Weight 171.17
Percent Purity 96%
Chemical Name or Material 2,4-Difluorophenyl isothiocyanate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.