missing translation for 'onlineSavingsMsg'
Learn More

2,4-Difluorophenylboronic acid, 98%

Product Code. p-2697092 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
1g
50g
This item is not returnable. View return policy

Product Code. 10521292

Brand: Thermo Scientific Acros 359130010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 144025-03-6 | C6H5BF2O2 | 157.91 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 144025-03-6
Molecular Formula C6H5BF2O2
Molecular Weight (g/mol) 157.91
MDL Number MFCD01318998
InChI Key QQLRSCZSKQTFGY-UHFFFAOYSA-N
Synonym 2,4-difluorobenzeneboronic acid, 2,4-difluorophenyl boronic acid, 2,4-fluorobenzene boronic acid, 2,4-difluorophenyl boranediol, 2,4-difluorophenyl-boronic acid, 2,4-difluorophenylboronicacid, 2 4-difluorophenylboronic acid, boronic acid, 2,4-difluorophenyl, boronic acid, b-2,4-difluorophenyl
PubChem CID 2734334
IUPAC Name (2,4-difluorophenyl)boronic acid
SMILES OB(O)C1=CC=C(F)C=C1F

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 247.0°C to 250.0°C
Color Beige to White or Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: .. Other solubilities: soluble in ether,thf,dmso,dmf,methanol
Formula Weight 157.91
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2, 4-Difluorophenylboronic acid
Show More Show Less
compliance-icons
Product Identifier
  • 2,4-Difluorophenylboronic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.