missing translation for 'onlineSavingsMsg'
Learn More

2,4-Difluorophenylhydrazine hydrochloride, 97%

Product Code. 11444330
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11444330 5 g 5g
11458970 25 g 25g
11468970 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11444330 Supplier Thermo Scientific Alfa Aesar Supplier No. A11625.06
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 51523-79-6 | C6H6F2N2 | 144.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51523-79-6
Molecular Formula C6H6F2N2
Molecular Weight (g/mol) 144.13
MDL Number MFCD00013383
InChI Key RLZUIPTYDYCNQI-UHFFFAOYSA-N
Synonym 2,4-difluorophenylhydrazine hydrochloride, 2,4-difluorophenyl hydrazine hydrochloride, 2,4-difluorophenylhydrazine hcl, 2,4-difluorohydrazinobenzene hydrochloride, 1-2,4-difluorophenyl hydrazine hydrochloride, 2,4-difluorophenylhydrazinehydrochloride, hydrazine, 2,4-difluorophenyl-, monohydrochloride, 2,4-difluorophenylhydrazine, chloride, pubchem3498, pubchem20774
PubChem CID 2733301
IUPAC Name (2,4-difluorophenyl)hydrazine;hydrochloride
SMILES NNC1=CC=C(F)C=C1F

Specifications

Melting Point ∼255°C (decomposition)
Quantity 5 g
Beilstein 5411448
Formula Weight 180.59
Percent Purity 98%
Chemical Name or Material 2,4-Difluorophenylhydrazine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.