missing translation for 'onlineSavingsMsg'
Learn More

2',4'-Dihydroxy-2-benzoylbenzoic Acid, TRC

Product Code. 30136566
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136566 10 g 10g
17961745 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30136566 Supplier Toronto Research Chemicals Supplier No. TRCD45175010G
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2513-33-9
Molecular Formula C14 H10 O5
Molecular Weight (g/mol) 258.23
Synonym 2-(2,4-Dihydroxybenzoyl)benzoic Acid, Fluorescein Sodium Imp. C (EP)
IUPAC Name 2-(2,4-dihydroxybenzoyl)benzoic acid
SMILES OC(=O)c1ccccc1C(=O)c2ccc(O)cc2O

Specifications

Quantity 10 g
InChI Formula InChI=1S/C14H10O5/c15-8-5-6-11(12(16)7-8)13(17)9-3-1-2-4-10(9)14(18)19/h1-7,15-16H,(H,18,19)
Formula Weight 258.0528
Percent Purity >95%
Chemical Name or Material 2-(2,4-Dihydroxybenzoyl)benzoic Acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.