missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dihydroxybenzoic acid, 97%

Product Code. 11480024
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11480024 2500 g 2500g
11460024 100 g 100g
11470024 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11480024 Supplier Thermo Scientific Alfa Aesar Supplier No. A13545.0E
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 89-86-1 | C7H6O4 | 154.121 g/mol

2,4-Dihydroxybenzoic acid is used in Intermediates, Plasticizers, Plastic and Rubber Products, a organic building block.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4-Dihydroxybenzoic acid is used in Intermediates, Plasticizers, Plastic and Rubber Products, a organic building block.

Solubility
Soluble in water(70g/L).

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89-86-1
Molecular Formula C7H6O4
Molecular Weight (g/mol) 154.121
MDL Number MFCD00002451
InChI Key UIAFKZKHHVMJGS-UHFFFAOYSA-N
Synonym beta-resorcylic acid, p-hydroxysalicylic acid, 4-carboxyresorcinol, 4-hydroxysalicylic acid, benzoic acid, 2,4-dihydroxy, beta-resorcinolic acid, 2,4-dhba, resorcylic acid, beta, b-resorcylic acid, 2,4-dihydroxy-benzoic acid
PubChem CID 1491
IUPAC Name 2,4-dihydroxybenzoic acid
SMILES C1=CC(=C(C=C1O)O)C(=O)O

Specifications

Melting Point ∼226°C (decomposition)
Quantity 2500 g
Beilstein 1946213
Merck Index 14,8157
Solubility Information Soluble in water(70g/L).
Formula Weight 154.12
Percent Purity 97%
Chemical Name or Material 2,4-Dihydroxybenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.