missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethoxybenzoic acid, 98%

Product Code. p-7023151
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11407717

Brand: Thermo Scientific Alfa Aesar A16252.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 91-52-1 | C9H10O4 | 182.175 g/mol

Yttrium and heavy lanthanide complexes is synthesised with 2, 4-dimethoxybenzoic acid and thermal and spectral features is studied. In the case of the electron-rich aromatic system 2,4-dimethoxybenzoic acid reduction of the intermediate HOBt ester is slow, and good.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Yttrium and heavy lanthanide complexes is synthesised with 2, 4-dimethoxybenzoic acid and thermal and spectral features is studied. In the case of the electron-rich aromatic system 2,4-dimethoxybenzoic acid reduction of the intermediate HOBt ester is slow, and good.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91-52-1
Molecular Formula C9H10O4
Molecular Weight (g/mol) 182.175
MDL Number MFCD00002434
InChI Key GPVDHNVGGIAOQT-UHFFFAOYSA-N
Synonym benzoic acid, 2,4-dimethoxy, 2,4-dimethoxybenzoicacid, 2,4-dimethoxy benzoic acid, 2,4-dimethoxybenzoic, pubchem4949, rarechem al bo 0028, 2,4-dimethoxy-benzoicaci, 2,4 dimethoxybenzoic acid, 2.4-dimethoxybenzoic acid, dimethoxybenzoic-2,4 acid
PubChem CID 7052
IUPAC Name 2,4-dimethoxybenzoic acid
SMILES COC1=CC(=C(C=C1)C(=O)O)OC

Specifications

Melting Point 107°C to 111°C
Odor Odorless
Quantity 25 g
Beilstein 2210860
Solubility Information Slightly soluble in water.
Formula Weight 182.18
Percent Purity 98%
Chemical Name or Material 2,4-Dimethoxybenzoic acid
compliance-icons
Product Identifier
  • 2,4-Dimethoxybenzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : 2,4-Dimethoxybenzoicacid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.