missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethoxybenzoic Acid 99%, ACROS Organics™

Product Code. 10457870 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
25g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10457870 25g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10457870 Supplier Thermo Scientific Acros Supplier No. 115430250

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 91-52-1
CAS Min % 96.0
CAS Max % 100.0
Melting Point 107°C to 111°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C9H10O4
Linear Formula (CH3O)2C6H3CO2H
MDL Number MFCD00002434
Quantity 25 g
Synonym benzoic acid, 2,4-dimethoxy, 2,4-dimethoxybenzoicacid, 2,4-dimethoxy benzoic acid, 2,4-dimethoxybenzoic, pubchem4949, rarechem al bo 0028, 2,4-dimethoxy-benzoicaci, 2,4 dimethoxybenzoic acid, 2.4-dimethoxybenzoic acid, dimethoxybenzoic-2,4 acid
InChI Key GPVDHNVGGIAOQT-UHFFFAOYSA-N
SMILES COC1=CC(=C(C=C1)C(=O)O)OC
IUPAC Name 2,4-dimethoxybenzoic acid
Molecular Weight (g/mol) 182.175
PubChem CID 7052
Formula Weight 182.18
Percent Purity ≥97.5%
Physical Form Powder
Chemical Name or Material 2, 4-Dimethoxybenzoic acid, 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.