missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethyl-2-pentenoic acid, predominantly trans, 99%

Product Code. 11455347
Change view
Click to view available options
Quantity:
2.5 g
10 g
Unit Size:
10g
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11455347 2.5 g 2.5g
11465347 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11455347 Supplier Thermo Scientific Alfa Aesar Supplier No. B22701.88
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 66634-97-7 | C7H12O2 | 128.171 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66634-97-7
Molecular Formula C7H12O2
Molecular Weight (g/mol) 128.171
MDL Number MFCD00209505
InChI Key DMHLGGQHOSTMJG-GQCTYLIASA-N
Synonym 2,4-dimethyl-2-pentenoic acid, unii-ki18c46dhr, 2e-2,4-dimethylpent-2-enoic acid, ki18c46dhr, 2,4-dimethyl-2-pentenoic acid, 2e, e-2,4-dimethyl-2-pentenoic acid, 2,4-dimethyl-2-pentenoic acid, predominantly trans, fema no. 3143, e-2,4-dimethylpent-2-enoic acid, 2-pentenoic acid, 2,4-dimethyl
PubChem CID 5362565
IUPAC Name (E)-2,4-dimethylpent-2-enoic acid
SMILES CC(C)C=C(C)C(=O)O

Specifications

Density 0.949
Refractive Index 1.454
Quantity 2.5 g
UN Number UN3265
Formula Weight 128.17
Percent Purity 99%
Chemical Name or Material 2,4-Dimethyl-2-pentenoic acid, predominantly trans

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.