missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethylthiophenol, 95%

Product Code. 11446396
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11446396 5 g 5g
11450747 25 g 25g
11460747 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11446396 Supplier Thermo Scientific Alfa Aesar Supplier No. B23161.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13616-82-5 | C8H10S | 138.228 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13616-82-5
Molecular Formula C8H10S
Molecular Weight (g/mol) 138.228
MDL Number MFCD00010019
InChI Key AMNLXDDJGGTIPL-UHFFFAOYSA-N
Synonym 2,4-dimethylthiophenol, 2,4-dimethyl thiophenol, benzenethiol, 2,4-dimethyl, 2,4-thioxylenol, 2,4-xylenethiol, 4-mercapto-m-xylene, 2,4-dimethylbenzene-1-thiol, 4-methylthiocresol, 4-thio-m-xylene, m-xylene-4-thiol
PubChem CID 83617
IUPAC Name 2,4-dimethylbenzenethiol
SMILES CC1=CC(=C(C=C1)S)C

Specifications

Density 1.02
Boiling Point 207°C to 208°C
Flash Point 87°C (189°F)
Odor Unpleasant
Refractive Index 1.569
Quantity 5 g
UN Number UN2810
Sensitivity Air Sensitive
Formula Weight 138.23
Percent Purity 95%
Chemical Name or Material 2,4-Dimethylthiophenol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2,4-Dimethylthiophenol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 2,4-Dimethylthiophenol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.