missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dinitroaniline, 99%

Product Code. 10486910 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10486910 5 g Glass bottle 5g
10316560 100 g Glass bottle 100g
10295100 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 10486910 Supplier Thermo Scientific Acros Supplier No. 116930050
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 97-02-9 | C6H5N3O4 | 183.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97-02-9
Molecular Formula C6H5N3O4
Molecular Weight (g/mol) 183.12
MDL Number MFCD00007151
InChI Key LXQOQPGNCGEELI-UHFFFAOYSA-N
Synonym benzenamine, 2,4-dinitro, 1-amino-2,4-dinitrobenzene, 2,4-dinitroanilin, 2,4-dinitrobenzenamine, 2,4-dinitroanilina, 2,4-dinitraniline, 2,4-dinitrophenylamine, aniline, 2,4-dinitro, 2,4-dinitroaminobenzene, 2,4-nitroaniline
PubChem CID 7321
ChEBI CHEBI:34242
IUPAC Name 2,4-dinitroaniline
SMILES NC1=CC=C(C=C1[N+]([O-])=O)[N+]([O-])=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 177°C to 180°C
Color Brown-Yellow or Green-Yellow to Yellow
Flash Point 224°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Linear Formula (O2N)2C6H3NH2
Quantity 5 g
Beilstein 12,747
Merck Index 15,3296
Solubility Information Solubility in water: 0.06g/L (20 c). Other solubilities: 0.7% ethanol (21 c),soluble in diethyl ether
Formula Weight 183.12
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 2, 4-Dinitroaniline, 99%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2,4-Dinitroaniline
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H310-Fatal in contact with skin.
  • H330-Fatal if inhaled.
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
  • H300-Fatal if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P361-Take off immediately all contaminated clothing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.