missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dinitrobenzaldehyde, 98%

Product Code. 11472807
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11472807

Brand: Thermo Scientific Alfa Aesar A15452.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 528-75-6 | C7H4N2O5 | 196.118 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 528-75-6
Molecular Formula C7H4N2O5
Molecular Weight (g/mol) 196.118
MDL Number MFCD00013376
InChI Key ZILXIZUBLXVYPI-UHFFFAOYSA-N
Synonym benzaldehyde, 2,4-dinitro, unii-zac919yu3z, ccris 2684, zac919yu3z, pubchem8223, benzaldehyde,4-dinitro, 2,4-dinitro-benzaldehyde, acmc-209l2f, 3-07-00-00923 beilstein handbook reference, ksc490k9l
PubChem CID 68250
IUPAC Name 2,4-dinitrobenzaldehyde
SMILES C1=CC(=C(C=C1[N+](=O)[O-])[N+](=O)[O-])C=O

Specifications

Melting Point 65°C to 68°C
Boiling Point 190°C (10 mmHg)
Quantity 25 g
Beilstein 1878706
Sensitivity Air Sensitive
Merck Index 14,3272
Formula Weight 196.12
Percent Purity 98%
Chemical Name or Material 2,4-Dinitrobenzaldehyde

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.