missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dinitrobenzenesulfonic acid hydrate, 98%

Product Code. 11459407
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11459407 100 g 100g
11449407 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11459407 Supplier Thermo Scientific Alfa Aesar Supplier No. B21430.22
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 698999-22-3 | C6H6N2O8S | 266.18 g/mol

2,4-Dinitrobenzenesulfonic acid hydrate is used to prepare ether-soluble 2,4-dinitrophenyl derivatives of amino alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4-Dinitrobenzenesulfonic acid hydrate is used to prepare ether-soluble 2,4-dinitrophenyl derivatives of amino alcohols.

Solubility
Soluble in methanol (2.5% w/v), slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under normal temperatures and pressures. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 698999-22-3
Molecular Formula C6H6N2O8S
Molecular Weight (g/mol) 266.18
MDL Number MFCD00150961
InChI Key GWGBNENHEGYJSN-UHFFFAOYSA-N
Synonym 2,4-dinitrobenzenesulfonic acid hydrate, dinitrobenzenesulfonic acid hydrate, c6h4n2o7s.h2o
PubChem CID 16217154
IUPAC Name 2,4-dinitrobenzenesulfonic acid;hydrate
SMILES C1=CC(=C(C=C1[N+](=O)[O-])[N+](=O)[O-])S(=O)(=O)O.O

Specifications

Melting Point 107°C to 109°C
Quantity 100 g
UN Number UN2585
Solubility Information Soluble in methanol (2.5% w/v),slightly soluble in water.
Formula Weight 248.17 (Anhydrous)
Percent Purity 98%
Chemical Name or Material 2,4-Dinitrobenzenesulfonic acid hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.