missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dinitrobenzenesulfonic acid sodium salt, 97%

Product Code. 11428407
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11428407 5 g 5g
11438407 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11428407 Supplier Thermo Scientific Alfa Aesar Supplier No. A18493.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 885-62-1 | C6H3N2NaO7S | 270.15 g/mol

2,4-Dinitrobenzenesulfonic acid sodium salt is used in the synthesis of aromatic dimethylene spacer-inserted cyclodextrin derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4-Dinitrobenzenesulfonic acid sodium salt is used in the synthesis of aromatic dimethylene spacer-inserted cyclodextrin derivatives.

Solubility
Soluble in water and methanol.

Notes
Hygroscopic. Moisture sensitive. Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 885-62-1
Molecular Formula C6H3N2NaO7S
Molecular Weight (g/mol) 270.15
MDL Number MFCD00007471,MFCD10567393
InChI Key GSBYVRKLPCSLNV-UHFFFAOYSA-M
Synonym sodium 2,4-dinitrobenzenesulfonate hydrate, acmc-209quh, sodium dnbs hydrate, 2,4-dinitrobenzenesulfonic acid sodium salt hydrate
PubChem CID 70700115
IUPAC Name sodium;2,4-dinitrobenzenesulfonate;hydrate
SMILES [Na+].[O-][N+](=O)C1=CC=C(C(=C1)[N+]([O-])=O)S([O-])(=O)=O

Specifications

Quantity 5 g
Beilstein 6676807
Solubility Information Soluble in water and methanol.
Formula Weight 270.15
Percent Purity 97%
Chemical Name or Material 2,4-Dinitrobenzenesulfonic acid sodium salt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.