missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dinitrobenzoic acid, 98%

Product Code. 11491898
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11491898 25 g 25g
11481898 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11491898 Supplier Thermo Scientific Alfa Aesar Supplier No. A14536.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 610-30-0 | C7H4N2O6 | 212.117 g/mol

2,4-Dinitrobenzoic acid was used in the spectrophotometric determination of diazepam in pure samples and in its pharmaceutical preparations. It was employed as chromophore probe for analysis of perfluorinated carboxylic acids in water by capillary zone electrophoresis. It can be used to produce 2,4-dinitro-benzoyl chloride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4-Dinitrobenzoic acid was used in the spectrophotometric determination of diazepam in pure samples and in its pharmaceutical preparations. It was employed as chromophore probe for analysis of perfluorinated carboxylic acids in water by capillary zone electrophoresis. It can be used to produce 2,4-dinitro-benzoyl chloride.

Solubility
Soluble in dioxane 50 mg/ml.

Notes
Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 610-30-0
Molecular Formula C7H4N2O6
Molecular Weight (g/mol) 212.117
MDL Number MFCD00017006
InChI Key ZIIGSRYPZWDGBT-UHFFFAOYSA-N
Synonym 2,4-dinitrobenzoic acid, benzoic acid, 2,4-dinitro, ccris 1766, acmc-20aoc2, 2.4-dinitrobenzoic acid, benzoicacid, 2,4-dinitro, 2,4-dinitrobenzoic acidanion, 4-09-00-01239 beilstein handbook reference, aronis26960, 1-carboxy-2,4-dinitrobenzene
PubChem CID 11880
SMILES C1=CC(=C(C=C1[N+](=O)[O-])[N+](=O)[O-])C(=O)O

Specifications

Melting Point 178°C to 184°C
Quantity 25 g
Beilstein 658650
Solubility Information Soluble in dioxane 50mg/ml.
Formula Weight 212.12
Percent Purity 98%
Chemical Name or Material 2,4-Dinitrobenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.